Alfa Aesar™ cis-1,2-Dimethylcyclohexane, 99+%


Overview
Brand: Alfa Aesar™

Manufacturer Part Number: 031467.09

UNSPSC: 12352001

Code: NEW

Additional Details:
Additional Details: CAS Number: 2207-01-4 Weight: 0.01000kg


Quantity 10g

Product Code. 11387468

Quantity Price
1 £ 46.43 / 10g
Estimated Shipment date
from Supplier 13-03-2017
Add to basket


Description and Specification

Chemical Identifiers

Chemical Name or Material cis-1,2-Dimethylcyclohexane
CAS 2207-01-4
Formula Weight 112.22
MDL Number MFCD00063621
UN Number UN2263

Specification

Formula Weight 112.22
Health Hazard 3 GHS P Statement
P210-P280-P240-P303+P361+P353-P403+P235-P501a
Keep away from heat/sparks/open flames/hot surfaces.
No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
Ground/bond container and receiving equipment.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing.
Rinse skin with water/shower.
Store in a well-ventilated place.
Keep cool.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Physical Form Liquid
Flash Point 12° (54°F)
MDL Number MFCD00063621
Health Hazard 2 GHS H Statement
H225
Highly flammable liquid and vapor.
Refractive Index 1.4360
CAS 2207-01-4
Assay ≥99%
DOT Information Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: DIMETHYLCYCLOHEXANES
Health Hazard 1 Danger
Boiling Point 129° to 130°C
Quantity 10g
Chemical Name or Material cis-1,2-Dimethylcyclohexane
Density 0.796g/mL
TSCA TSCA
UN Number UN2263
Safety and Handling

Danger

GHS H Statement
H225
Highly flammable liquid and vapor.

GHS P Statement
P210-P280-P240-P303+P361+P353-P403+P235-P501a
Keep away from heat/sparks/open flames/hot surfaces.
No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
Ground/bond container and receiving equipment.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing.
Rinse skin with water/shower.
Store in a well-ventilated place.
Keep cool.
Dispose of contents/container in accordance with local/regional/national/international regulations.