missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ cis-1,2-Dimethylcyclohexane, 99+%

Brand:  Alfa Aesar™ 031467.18

Code : NEW

Additional Details : CAS Number : 2207-01-4 Weight : 0.05000kg Transport : UN number : 2263 Chem class : 3 Pack group : II

Quantity 50g

 View more versions of this product

Product Code. 11377468

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Product Support team on 01509 555888 for assistance


Specifications

Chemical Identifiers

cis-1,2-Dimethylcyclohexane
112.22
UN2263
2207-01-4
MFCD00063621

Specifications

cis-1,2-Dimethylcyclohexane
0.796g/mL
≥99%
MFCD00063621
1.4360
50g
2207-01-4
Liquid
112.22
UN2263
129° to 130°C
12° (54°F)
Videos
SDS