missing translation for 'onlineSavingsMsg'
Learn More

cis-1,2-Dichloroethylene, 97%, stabilized, ACROS Organics™

Brand:  Acros Organics 113380500

Code : 29

Additional Details : CAS Number : 156-59-2 Weight : 0.11400kg Transport : UN number : 1150 Chem class : 3 Pack group : II

Quantity 50mL
Packaging Glass bottle

 View more versions of this product

Product Code. 10452865

  • £378.00 / 50mL


Estimated Shipment
19-06-2018
Add to basket

Specifications

Chemical Identifiers

cis-1, 2-Dichloroethylene
156-59-2
97%
ClCH=CHCl
MFCD00000925
97%
97%
C2H2Cl2
96.94

Specifications

cis-1, 2-Dichloroethylene
6°C
60.0°C
− 80.0°C
97%
Authentic
1.4475 to 1.4495
Liquid
97%
ClCH=CHCl
MFCD00000925
1.2800g/mL
1.28
Clear colorless
50mL
156-59-2
Glass bottle
0.003% MEHQ
97%
C2H2Cl2
96.94
Videos
SDS