missing translation for 'onlineSavingsMsg'
Learn More

Cinnamonitrile, 97%, predominantly trans, Thermo Scientific Chemicals

Catalog Number p-7028279
Click to view available options
Quantity:
25 g
100 g
500 g
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21615” to get your promotional price
1885-38-7
C9H7N
129.162
MFCD00001930
ZWKNLRXFUTWSOY-QPJJXVBHSA-N
cinnamonitrile, trans-cinnamonitrile, styryl cyanide, cinnamyl nitrile, 3-phenylacrylonitrile, beta-cyanostyrene, cinnamonitrile, e, 3-phenyl-2-propenenitrile, acrylonitrile, 3-phenyl, e-3-phenylprop-2-enenitrile
1550846
(E)-3-phenylprop-2-enenitrile
C1=CC=C(C=C1)C=CC#N
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21615” to get your promotional price
1885-38-7
C9H7N
129.162
MFCD00001930
ZWKNLRXFUTWSOY-QPJJXVBHSA-N
cinnamonitrile, trans-cinnamonitrile, styryl cyanide, cinnamyl nitrile, 3-phenylacrylonitrile, beta-cyanostyrene, cinnamonitrile, e, 3-phenyl-2-propenenitrile, acrylonitrile, 3-phenyl, e-3-phenylprop-2-enenitrile
1550846
(E)-3-phenylprop-2-enenitrile
C1=CC=C(C=C1)C=CC#N

Cinnamonitrile is a parent compound whose derivatives can be applied as corrosion inhibitors for API J55 steel.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Cinnamonitrile is a parent compound whose derivatives can be applied as corrosion inhibitors for API J55 steel.

Solubility
Soluble in alcohol. Insoluble in water.

Notes
Air & light sensitive. Store in dark. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY
Melting Point 18°C to 20°C
Density 1.029
Boiling Point 253°C to 254°C
Flash Point 126°C (259°F)
Refractive Index 1.602
Quantity 25 g
Sensitivity Air and light sensitive
Solubility Information Soluble in alcohol. Insoluble in water.
Formula Weight 129.16
Percent Purity 97%
Chemical Name or Material Cinnamonitrile, predominantly trans
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'