missing translation for 'onlineSavingsMsg'
Learn More

Cinnabarinic acid, Tocris Bioscience™

Selective mGlu4 agonist

Brand:  Tocris (Bio-Techne) 411910

Code : Z1

Additional Details : Weight : 0.01000kg

Quantity 10mg

 View more versions of this product

Product Code. 13464184

  • £170.28 / 10mg


Estimated Shipment
05-04-2019
Add to basket

Description

Description

mGlu 4 receptor agonist. Activates mGlu 4 receptors in transiently transfected HEK293 cells; selective over all other mGlu subtypes (mGlu 1 mGlu 2 and mGlu 5-8 ). Decreases cAMP levels in native cultured cerebellar granule cells. Also counteracts excitotoxic neuronal cell death induced by NMDA in mixed cultures of cortical cells; displays neuroprotective activity.
Specifications

Chemical Identifiers

606-59-7
300.226
cinnabarinic acid, cinnavalininate, 2-amino-3-oxo-3h-phenoxazine-1,9-dicarboxylic acid, cinnabaric acid, 2-amino-3h-phenoxazin-one-1,9-dicarboxylic acid, 2-amino-3-oxo-3h-phenoxazin-1,9-dicarboxylic acid, 3h-phenoxazine-1,9-dicarboxylicacid, 2-amino-3-oxo, cinnabarinic acid/, cinnavalininic acid, cinnabarinic acid hplc
CHEBI:3715
C1=CC(=C2C(=C1)OC3=CC(=O)C(=C(C3=N2)C(=O)O)N)C(=O)O
C14H8N2O6
FSBKJYLVDRVPTK-UHFFFAOYSA-N
114918
2-amino-3-oxophenoxazine-1,9-dicarboxylic acid

Specifications

Cinnabarinic acid
606-59-7
cinnabarinic acid, cinnavalininate, 2-amino-3-oxo-3h-phenoxazine-1,9-dicarboxylic acid, cinnabaric acid, 2-amino-3h-phenoxazin-one-1,9-dicarboxylic acid, 2-amino-3-oxo-3h-phenoxazin-1,9-dicarboxylic acid, 3h-phenoxazine-1,9-dicarboxylicacid, 2-amino-3-oxo, cinnabarinic acid/, cinnavalininic acid, cinnabarinic acid hplc
C1=CC(=C2C(=C1)OC3=CC(=O)C(=C(C3=N2)C(=O)O)N)C(=O)O
300.226
CHEBI:3715
>98%
10mg
C14H8N2O6
FSBKJYLVDRVPTK-UHFFFAOYSA-N
2-amino-3-oxophenoxazine-1,9-dicarboxylic acid
114918
300.22
Videos