missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific Chemicals Cinchonidine, 98.5-101%
39.65 GBP valid until 2024-12-31
Use promo code "22914" to get your promotional price.
Use promo code "22914" to get your promotional price.
Click to view available options
Quantity:
25 g
100 g
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
General Description
Cinchonidine is an alkaloid naturally occurring in the bark of Cinchona strubs that has activity against Plasmodium spp
Cinchonidine is a diastereoisomer of cinchonine and can be used to promote chirality in asymmetric or enantioselective synthesis of organic compounds
Applications
Cinchonidine has been used as a starting material in the synthesis of anti-malarial cinchona alkaloid conjugates
Has been shown to potently induce γ-globin mRNA and fetal hemoglobin in erythroid progenitor cells and may be useful in preclinical studies of β-thalassemia
Specifications
Melting Point | 200.0°C to 203.0°C |
Color | White to Cream |
Loss on Drying | 1% max. (105°C) |
Infrared Spectrum | Authentic |
Assay Percent Range | 98.5-101% |
Packaging | Glass bottle |
Quantity | 25 g |
Beilstein | 23, 420 |
Merck Index | 15, 2289 |
Specific Rotation | − 115.00 |
Show More |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'