missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cinchocaine Hydrochloride EP Impurity D, Mikromol™
Click to view available options
Quantity:
25 mg
100 mg
Unit Size:
100mg
25mg
Description
Cinchocaine Hydrochloride EP Impurity D
Specifications
Specifications
| Chemical Name or Material | 2-Butoxyquinoline-4-carboxylicAcid |
| CAS | 10222-61-4 |
| Analyte or Component Names | 2-Butoxyquinoline-4-carboxylicAcid |
| Shipping Condition | Room Temperature |
| Molecular Formula | C14 H15 N O3 |
| Certifications/Compliance | ISO 17025 |
| Synonym | 2-Butoxyquinoline-4-carboxylic Acid, Cinchocaine Hydrochloride Imp. D (EP), Cinchocaine Imp. D (EP), 4-Quinolinecarboxylic acid, 2-butoxy-, Cinchoninic acid, 2-butoxy- (7CI,8CI), 2-Butoxy-4-quinolinecarboxylic acid, Cinchocaine Hydrochloride Impurity D, Cinchocaine Impurity D |
| InChI Formula | InChI=1S/C14H15NO3/c1-2-3-8-18-13-9-11(14(16)17)10-6-4-5-7-12(10)15-13/h4-7,9H,2-3,8H2,1H3,(H,16,17) |
| SMILES | O=C(O)C=1C=C(N=C2C=CC=CC21)OCCCC |
| IUPAC Name | 2-butoxyquinoline-4-carboxylic acid |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction