Learn More
Cinanserin hydrochloride, Tocris Bioscience™
Selective 5-HT2 antagonist
Brand:  Tocris (Bio-Techne) 046050
Code : Z1
Additional Details : Weight : 0.01000kg
Quantity 50mg
 View more versions of this product
Product Code. 13447673
  • £448.35 / 50mg
Estimated Shipment date
from Supplier 21-11-2019
Description
Description
5-HT2 antagonist.
Specifications
Chemical Identifiers
54-84-2
376.943
cinanserin hydrochloride, cinnanserin, maptc, cinanserin hcl, unii-f65z32ki62, cinanserin hydrochloride usan, 2'-3-dimethylamino propyl thio cinnamanilide hydrochloride, 2'-3-dimethylamino propyl thio cinnamanilide monohydrochloride, 2-propenamide, n-2-3-dimethylamino propyl thio phenyl-3-phenyl-, monohydrochloride
(E)-N-[2-[3-(dimethylamino)propylsulfanyl]phenyl]-3-phenylprop-2-enamide;hydrochloride
C20H25ClN2OS
LXGJPDKYMJJWRB-IERUDJENSA-N
6433141
CN(C)CCCSC1=CC=CC=C1NC(=O)C=CC2=CC=CC=C2.Cl
Specifications
50mg
54-84-2
cinanserin hydrochloride, cinnanserin, maptc, cinanserin hcl, unii-f65z32ki62, cinanserin hydrochloride usan, 2'-3-dimethylamino propyl thio cinnamanilide hydrochloride, 2'-3-dimethylamino propyl thio cinnamanilide monohydrochloride, 2-propenamide, n-2-3-dimethylamino propyl thio phenyl-3-phenyl-, monohydrochloride
CN(C)CCCSC1=CC=CC=C1NC(=O)C=CC2=CC=CC=C2.Cl
376.943
376.94
Cinanserin hydrochloride
C20H25ClN2OS
LXGJPDKYMJJWRB-IERUDJENSA-N
(E)-N-[2-[3-(dimethylamino)propylsulfanyl]phenyl]-3-phenylprop-2-enamide;hydrochloride
6433141
>99%