missing translation for 'onlineSavingsMsg'
Learn More

Cinalukast, Tocris Bioscience™

Potent, selective CysLT1 (LTD4) antagonist; orally active

Brand:  Tocris (Bio-Techne) 202550

Code : Z1

Additional Details : Weight : 0.01000kg

Quantity 50mg

 View more versions of this product

Product Code. 13459383

  • £600.29 / 50mg


Estimated Shipment
07-03-2019
Add to basket

Description

Description

Potent, selective CysLT1 (LTD4) leukotriene receptor antagonist (IC50 = 6.4 nM). Inhibits LTD4-induced bronchoconstriction in guinea pigs when administered intravenously, orally or by aerosol.
Specifications

Chemical Identifiers

128312-51-6
412.548
cinalukast, cinalukast usan:inn, unii-5e1o433qai, cinalukast usan/inn, 3'-e-2-4-cyclobutyl-2-thiazolyl vinyl-2,2-diethylsuccinanilic acid, dsstox_cid_25742, dsstox_rid_81087, dsstox_gsid_45742, e-4-3-2-4-cyclobutyl-2-thiazolyl ethenyl phenyl amino-2,2-diethyl-4-oxobutanoic acid, 4-3-e-2-4-cyclobutyl-1,3-thiazol-2-yl ethenyl anilino-2,2-diethyl-4-oxobutanoic acid
CHEBI:126598
CCC(CC)(CC(=O)NC1=CC=CC(=C1)C=CC2=NC(=CS2)C3CCC3)C(=O)O
C23H28N2O3S
BZMKNPGKXJAIDV-VAWYXSNFSA-N
6436135
4-[3-[(E)-2-(4-cyclobutyl-1,3-thiazol-2-yl)ethenyl]anilino]-2,2-diethyl-4-oxobutanoic acid

Specifications

Cinalukast
128312-51-6
cinalukast, cinalukast usan:inn, unii-5e1o433qai, cinalukast usan/inn, 3'-e-2-4-cyclobutyl-2-thiazolyl vinyl-2,2-diethylsuccinanilic acid, dsstox_cid_25742, dsstox_rid_81087, dsstox_gsid_45742, e-4-3-2-4-cyclobutyl-2-thiazolyl ethenyl phenyl amino-2,2-diethyl-4-oxobutanoic acid, 4-3-e-2-4-cyclobutyl-1,3-thiazol-2-yl ethenyl anilino-2,2-diethyl-4-oxobutanoic acid
CCC(CC)(CC(=O)NC1=CC=CC(=C1)C=CC2=NC(=CS2)C3CCC3)C(=O)O
412.548
CHEBI:126598
>98%
50mg
C23H28N2O3S
BZMKNPGKXJAIDV-VAWYXSNFSA-N
4-[3-[(E)-2-(4-cyclobutyl-1,3-thiazol-2-yl)ethenyl]anilino]-2,2-diethyl-4-oxobutanoic acid
6436135
412.55
Videos