Learn More
Thermo Scientific™ Cimarec™ Digital Stirring Hotplates
Carry out general heating applications with Cimarec Digital Stirring Hotplates, which deliver precise stirring control, exceptional safety/temperature performance.
£388.28 - £460.62
Specifications
Type Hot Plate Stirrer, Stirrer
View More Specs
Products 3
Product Code Brand Size Voltage Top Plate Material Price Quantity & Availability  
Product Code Brand Size Voltage Top Plate Material Price Quantity & Availability  
11737228
View Documents Promotion Details
Thermo Scientific™
SP13132033Q
7.25 x 7.25 in. 220/240 V Ceramic This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
Save Up to 
»
11727228
View Documents Promotion Details
Thermo Scientific™
SP13101033Q
4.25 x 4.25 in. 220/240 V Ceramic This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
Save Up to 
»
10164673
View Documents Promotion Details
Thermo Scientific™
SP142020-33Q
7.25 x 7.25 in. 220/240 V Aluminum This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
Save Up to 
»
Description
Description
Carry out general heating applications with  Thermo Scientific™ Cimarec™ Digital Stirring Hotplates, which deliver precise stirring control, exceptional safety/temperature performance. They offer advanced stirring controls and are ideal for repetitive procedures demanding precision and safety.
Performance Features
  • Microprocessor-controlled feedback technology maintains consistent, repeatable temperature settings from 5°C up to maximum
  • Digital display and large control knob enable precise temperature control
  • Flat top and high-wattage heating elements provide superior heat transfer and fast time-to-boil
  • Various sizes offered to handle different sample volumes
  • Rugged cast aluminum body is stable and durable
  • StirTrac™ technology allows smooth low-speed stirring, consistent speed control and stronger magnetic coupling
  • StirTrac™ braking brings stir bar to immediate stop for quick flash removal
  • Integrated ring stand holder to accommodate 0.5™ diameter (1.3cm) support rod
Safety Features
  • Hot surface alert protects from accidental burns; light activates when heating surface is above 50°C (122°F)
  • Unique HOT TOP alert provides added protection
  • Unit displays HOT OFF until unit reaches 50°C (122°F) or below, even when heat control is turned off
  • Cast-aluminum base diverts spills from internal electronics
  • Low-profile design and stable, rugged base prevents tipping and spillage
Ceramic Top Models
  • Clean easily and resist alkalis and acids
  • Seamless, reflective white surface aids sample visibility
  • Heat to higher temperatures than aluminum tops
Aluminum Top Models
  • Rugged and chip-resistant for superior durability
  • Allow uniform heat distribution across surface
Includes:
Detachable line cord. All models supplied with a 0.38 dia. × 2 in. L (1 × 5.1cm) TFE-coated stir bar, except 4 x 4 in., which includes a 1 in. (2.54cm) stir bar.
Specifications
Specifications
Hot Plate Stirrer, Stirrer