missing translation for 'onlineSavingsMsg'
Learn More

Cilostamide, Tocris Bioscience™

PDE3 inhibitor

Brand:  Tocris (Bio-Techne) 091510

Code : Z1

Additional Details : Weight : 0.01000kg

Quantity 10mg

 View more versions of this product

Product Code. 13438153

  • £147.15 / 10mg

Estimated Shipment
27-06-2019

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Selective inhibitor of type III phosphodiesterase (PDE3). Displays moderate selectivity for PDE3A isozyme vs. PDE3B (IC50 values are 0.027 and 0.050 μM for PDE3A and PDE3B respectively). Inhibits ADP-induced platelet aggregation (IC50 = 16.8 μM); antithrombotic. Also available as part of the Phosphodiesterase Inhibitor Tocriset™ .
Specifications

Chemical Identifiers

68550-75-4
342.439
cilostamide, ciloalamide, cilostamide inn, cilostamidum inn-latin, cilostamida inn-spanish, n-cyclohexyl-n-methyl-4-1,2-dihydro-2-oxo-6-quinolyloxy butyramide, chembl34431, n-cyclohexyl-n-methyl-4-2-oxo-1h-quinolin-6-yl oxy butanamide, butanamide, n-cyclohexyl-4-1,2-dihydro-2-oxo-6-quinolinyl oxy-n-methyl, dsstox_cid_25140
N-cyclohexyl-N-methyl-4-[(2-oxo-1H-quinolin-6-yl)oxy]butanamide
C20H26N2O3
UIAYVIIHMORPSJ-UHFFFAOYSA-N
2753
CN(C1CCCCC1)C(=O)CCCOC2=CC3=C(C=C2)NC(=O)C=C3

Specifications

Cilostamide
68550-75-4
cilostamide, ciloalamide, cilostamide inn, cilostamidum inn-latin, cilostamida inn-spanish, n-cyclohexyl-n-methyl-4-1,2-dihydro-2-oxo-6-quinolyloxy butyramide, chembl34431, n-cyclohexyl-n-methyl-4-2-oxo-1h-quinolin-6-yl oxy butanamide, butanamide, n-cyclohexyl-4-1,2-dihydro-2-oxo-6-quinolinyl oxy-n-methyl, dsstox_cid_25140
CN(C1CCCCC1)C(=O)CCCOC2=CC3=C(C=C2)NC(=O)C=C3
342.439
342.44
10mg
C20H26N2O3
UIAYVIIHMORPSJ-UHFFFAOYSA-N
N-cyclohexyl-N-methyl-4-[(2-oxo-1H-quinolin-6-yl)oxy]butanamide
2753
>98%
Videos