Learn More
Cilastatin sodium, Tocris Bioscience™
Dipeptidase inhibitor
Brand:  Tocris (Bio-Techne) 270950
Code : Z1
Additional Details : Weight : 0.01000kg
Quantity 50mg
 View more versions of this product
Product Code. 13441774
  • £454.36 / 50mg
Estimated Shipment
11-09-2020
Description
Description
Dipeptidase inhibitor (LTDase, leukotriene D4 hydrolase, dehydropeptidase I) that displays a Ki value of 0.11 μM. Inhibits metabolism of LTD4 to LTE4 and the hydrolysis of β-lactam antibiotics. Nephroprotective; reduces toxic accumulation of cyclosporin A in kidney proximal tubule epithelial cells.
Specifications
Chemical Identifiers
81129-83-1
380.435
cilastatin sodium, cilastatin sodium salt
CHEBI:59511
CC1(CC1C(=NC(=CCCCCSCC(C(=O)O)N)C(=O)O)[O-])C.[Na+]
C16H25N2NaO5S
QXPBTTUOVWMPJN-QBNHLFMHSA-M
101282945
sodium;(1S)-N-[(Z)-6-[(2R)-2-amino-2-carboxyethyl]sulfanyl-1-carboxyhex-1-enyl]-2,2-dimethylcyclopropane-1-carboximidate
Specifications
Cilastatin sodium
81129-83-1
cilastatin sodium, cilastatin sodium salt
CC1(CC1C(=NC(=CCCCCSCC(C(=O)O)N)C(=O)O)[O-])C.[Na+]
380.435
CHEBI:59511
>99%
50mg
C16H25N2NaO5S
QXPBTTUOVWMPJN-QBNHLFMHSA-M
sodium;(1S)-N-[(Z)-6-[(2R)-2-amino-2-carboxyethyl]sulfanyl-1-carboxyhex-1-enyl]-2,2-dimethylcyclopropane-1-carboximidate
101282945
380.43