missing translation for 'onlineSavingsMsg'
Learn More

Ciglitazone, Tocris Bioscience™

Selective PPARγ agonist

Brand:  Tocris (Bio-Techne) 130710

Code : Z1

Additional Details : Weight : 0.01000kg

Quantity 10mg

 View more versions of this product

Product Code. 13439103

  • £175.56 / 10mg

Estimated Shipment
27-06-2019

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Selective agonist at PPARγ (peroxisome proliferator-activated receptor γ). Activates PPARγ with an EC50 value of 3 μM in vitro, and is at least 33-fold selective over PPARα and δ. Antihyperglycemic in vivo.
Specifications

Chemical Identifiers

74772-77-3
333.446
ciglitazone, ciglitizone, ciglitazona, ciglitazonum, ciglitazonum latin, ciglitazona spanish, ciglitazone usan:inn, +--5-p-1-methylcyclohexyl methoxy benzyl-2,4-thiazolidinedione, +/--5-4-1-methylcyclohexylmethoxy benzyl thiazolidine-2,4-dione, 5-4-1-methylcyclohexyl methoxy benzyl-1,3-thiazolidine-2,4-dione
CHEBI:64227
CC1(CCCCC1)COC2=CC=C(C=C2)CC3C(=O)NC(=O)S3
C18H23NO3S
YZFWTZACSRHJQD-UHFFFAOYSA-N
2750
5-[[4-[(1-methylcyclohexyl)methoxy]phenyl]methyl]-1,3-thiazolidine-2,4-dione

Specifications

Ciglitazone
74772-77-3
ciglitazone, ciglitizone, ciglitazona, ciglitazonum, ciglitazonum latin, ciglitazona spanish, ciglitazone usan:inn, +--5-p-1-methylcyclohexyl methoxy benzyl-2,4-thiazolidinedione, +/--5-4-1-methylcyclohexylmethoxy benzyl thiazolidine-2,4-dione, 5-4-1-methylcyclohexyl methoxy benzyl-1,3-thiazolidine-2,4-dione
CC1(CCCCC1)COC2=CC=C(C=C2)CC3C(=O)NC(=O)S3
333.446
CHEBI:64227
>99%
10mg
C18H23NO3S
YZFWTZACSRHJQD-UHFFFAOYSA-N
5-[[4-[(1-methylcyclohexyl)methoxy]phenyl]methyl]-1,3-thiazolidine-2,4-dione
2750
333.44
Videos