missing translation for 'onlineSavingsMsg'
Learn More

CID 2745687, Tocris Bioscience™

GPR35 antagonist

Brand:  Tocris (Bio-Techne) 429350

Code : Z1

Additional Details : Weight : 0.01000kg

Quantity 50mg

 View more versions of this product

Product Code. 13464504

  • £639.97 / 50mg


Estimated Shipment
03-04-2019
Add to basket

Description

Description

Competitive, reversible antagonist of the orphan receptor GPR35 (K i = 12.8 nM). Blocks the GPR35-mediated increase in ERK1/2 phosphorylation and β-arrestin recruitment induced by pamoic acid (Cat. No. 4298).
Specifications

Chemical Identifiers

264233-05-8
395.429
1-2,4-difluorophenyl-5-2-1,1-dimethylehyl amino thioxomethyl hydrazinylidene methyl-1h-pyrazole-4-carboxylic acid methyl ester, methyl 5-2-tert-butylamino carbothioyl carbohydrazonoyl-1-2,4-difluorophenyl-1h-pyrazole-4-carboxylate, ml hplc, 5-e-tert-butylthiocarbamoylhydrazono methyl-1-2,4-difluorophenyl pyrazole-4-carboxylic acid methyl ester, 5-tert-butylthiocarbamoylhydrazono methyl-1-2,4-difluorophenyl pyrazole-4-carboxylic acid methyl ester, 5-tert-butylamino-sulfanylidenemethyl hydrazinylidene methyl-1-2,4-difluorophenyl-4-pyrazolecarboxylic acid methyl ester, methyl 1-2,4-difluoro-phenyl-5-4-tert-butyl-thiosemicarbazono methyl-1h-pyrazole-4-carboxylate, methyl 1-2,4-bis fluoranyl phenyl-5-tert-butylcarbamothioylhydrazinylidene methyl pyrazole-4-carboxylate
methyl 5-[(E)-(tert-butylcarbamothioylhydrazinylidene)methyl]-1-(2,4-difluorophenyl)pyrazole-4-carboxylate
C17H19F2N5O2S
CYNLZIBKERMMOA-AWQFTUOYSA-N
9581011
CC(C)(C)NC(=S)NN=CC1=C(C=NN1C2=C(C=C(C=C2)F)F)C(=O)OC

Specifications

CID 2745687
264233-05-8
1-2,4-difluorophenyl-5-2-1,1-dimethylehyl amino thioxomethyl hydrazinylidene methyl-1h-pyrazole-4-carboxylic acid methyl ester, methyl 5-2-tert-butylamino carbothioyl carbohydrazonoyl-1-2,4-difluorophenyl-1h-pyrazole-4-carboxylate, ml hplc, 5-e-tert-butylthiocarbamoylhydrazono methyl-1-2,4-difluorophenyl pyrazole-4-carboxylic acid methyl ester, 5-tert-butylthiocarbamoylhydrazono methyl-1-2,4-difluorophenyl pyrazole-4-carboxylic acid methyl ester, 5-tert-butylamino-sulfanylidenemethyl hydrazinylidene methyl-1-2,4-difluorophenyl-4-pyrazolecarboxylic acid methyl ester, methyl 1-2,4-difluoro-phenyl-5-4-tert-butyl-thiosemicarbazono methyl-1h-pyrazole-4-carboxylate, methyl 1-2,4-bis fluoranyl phenyl-5-tert-butylcarbamothioylhydrazinylidene methyl pyrazole-4-carboxylate
CC(C)(C)NC(=S)NN=CC1=C(C=NN1C2=C(C=C(C=C2)F)F)C(=O)OC
395.429
395.43
50mg
C17H19F2N5O2S
CYNLZIBKERMMOA-AWQFTUOYSA-N
methyl 5-[(E)-(tert-butylcarbamothioylhydrazinylidene)methyl]-1-(2,4-difluorophenyl)pyrazole-4-carboxylate
9581011
>98%
Videos