missing translation for 'onlineSavingsMsg'
Learn More

CI 988, Tocris Bioscience™

Potent and selective CCK2 antagonist

Brand:  Tocris (Bio-Techne) 260710

Code : Z1

Additional Details : Weight : 0.01000kg

Quantity 10mg

 View more versions of this product

Product Code. 13441564

  • £252.13 / 10mg

Estimated Shipment
01-07-2019

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Potent and selective CCK2 (CCK-B) receptor antagonist that displays ∽ 1600-fold selectivity over CCK1 receptors (IC50 values are 1.7 and 2717 nM for CCK2 and CCK1 respectively). Has negligible affinity at a range of other binding sites (IC50 > 10 μM). Exhibits anxiolytic activity following oral administration.
Specifications

Chemical Identifiers

130332-27-3
614.743
unii-2637pdx9si, 3-1r-2-2r-2-2-adamantyloxycarbonylamino-3-1h-indol-3-yl-2-methyl-propanoyl amino-1-phenyl-ethyl carbamoyl propanoic acid, 4-1r-2-2r-2-2-adamantyloxycarbonylamino-3-1h-indol-3-yl-2-methylpropanoyl amino-1-phenylethyl amino-4-oxobutanoic acid, 4-1r-2-2r-3-1h-indol-3-yl-2-methyl-1-oxo-2-tricyclo 3.3.1.13,7 dec-2-yloxy carbonyl amino propyl amino-1-phenylethyl amino-4-oxobutanoic acid, gtpl873, butanoic acid, 4-1r-2-2r-3-1h-indol-3-yl-2-methyl-1-oxo-2-tricyclo 3.3.1.1 3,7 dec-2-yloxy carbonyl amino propyl amino-1-phenylethyl amino-4-oxo, r-r*,r*-4-2-3-1h-indol-3-yl-2-methyl-1-oxo-2-tricyclo 3.3.1.13,7 dec-2-yloxy carbonyl amino propyl amino-1-phenylethyl amino-4-oxobutanoic acid, 3-1r-2-2r-2-1h-indol-3-ylmethyl-2-1r,3r,5r,7r-adamantan-2-yloxy carbonyl amino propanamido-1-phenylethyl carbamoyl propanoic acid, 3-1r-2-2r-2-adamantan-2-yloxy carbonyl amino-2-1h-indol-3-ylmethyl propanamido-1-phenylethyl carbamoyl propanoic acid, 4-r-2-r-2-adamantan-2-yloxy carbonyl amino-3-1h-indol-3-yl-2-methylpropanamido-1-phenylethyl amino-4-oxobutanoic acid
4-[[(1R)-2-[[(2R)-2-(2-adamantyloxycarbonylamino)-3-(1H-indol-3-yl)-2-methylpropanoyl]amino]-1-phenylethyl]amino]-4-oxobutanoic acid
C35H42N4O6
FVQSSYMRZKLFDR-ZABPBAJSSA-N
108187
CC(CC1=CNC2=CC=CC=C21)(C(=O)NCC(C3=CC=CC=C3)NC(=O)CCC(=O)O)NC(=O)OC4C5CC6CC(C5)CC4C6

Specifications

CI 988
130332-27-3
unii-2637pdx9si, 3-1r-2-2r-2-2-adamantyloxycarbonylamino-3-1h-indol-3-yl-2-methyl-propanoyl amino-1-phenyl-ethyl carbamoyl propanoic acid, 4-1r-2-2r-2-2-adamantyloxycarbonylamino-3-1h-indol-3-yl-2-methylpropanoyl amino-1-phenylethyl amino-4-oxobutanoic acid, 4-1r-2-2r-3-1h-indol-3-yl-2-methyl-1-oxo-2-tricyclo 3.3.1.13,7 dec-2-yloxy carbonyl amino propyl amino-1-phenylethyl amino-4-oxobutanoic acid, gtpl873, butanoic acid, 4-1r-2-2r-3-1h-indol-3-yl-2-methyl-1-oxo-2-tricyclo 3.3.1.1 3,7 dec-2-yloxy carbonyl amino propyl amino-1-phenylethyl amino-4-oxo, r-r*,r*-4-2-3-1h-indol-3-yl-2-methyl-1-oxo-2-tricyclo 3.3.1.13,7 dec-2-yloxy carbonyl amino propyl amino-1-phenylethyl amino-4-oxobutanoic acid, 3-1r-2-2r-2-1h-indol-3-ylmethyl-2-1r,3r,5r,7r-adamantan-2-yloxy carbonyl amino propanamido-1-phenylethyl carbamoyl propanoic acid, 3-1r-2-2r-2-adamantan-2-yloxy carbonyl amino-2-1h-indol-3-ylmethyl propanamido-1-phenylethyl carbamoyl propanoic acid, 4-r-2-r-2-adamantan-2-yloxy carbonyl amino-3-1h-indol-3-yl-2-methylpropanamido-1-phenylethyl amino-4-oxobutanoic acid
CC(CC1=CNC2=CC=CC=C21)(C(=O)NCC(C3=CC=CC=C3)NC(=O)CCC(=O)O)NC(=O)OC4C5CC6CC(C5)CC4C6
614.743
614.73
10mg
C35H42N4O6
FVQSSYMRZKLFDR-ZABPBAJSSA-N
4-[[(1R)-2-[[(2R)-2-(2-adamantyloxycarbonylamino)-3-(1H-indol-3-yl)-2-methylpropanoyl]amino]-1-phenylethyl]amino]-4-oxobutanoic acid
108187
>99%
Videos