missing translation for 'onlineSavingsMsg'
Learn More

CI 1020, Tocris Bioscience™

Highly selective, orally active ETA antagonist

Brand:  Tocris (Bio-Techne) 294210

Code : Z1

Additional Details : Weight : 0.01000kg

Quantity 10mg

 View more versions of this product

Product Code. 13462094

  • £161.70 / 10mg

Estimated Shipment date
from Supplier 31-03-2020

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Highly selective, orally active, non-peptide endothelin-A receptor (ETA) antagonist (IC50 values are 0.3 and 480 nM for ETA and ETB receptors respectively). Antihypertensive; blocks ET-1-induced pressor responses following oral administration.
Specifications

Chemical Identifiers

162256-50-0
506.507
chembl8981, 3-1,3-benzodioxol-5-yl-5-hydroxy-5-4-methoxyphenyl-4-3,4,5-trimethoxyphenyl methyl-2 5h-furanone, 3-benzo 1,3 dioxol-5-yl-5-hydroxy-5-4-methoxy-phenyl-4-3,4,5-trimethoxy-benzyl-5h-furan-2-one, 2 5h-furanone,3-1,3-benzodioxol-5-yl-5-hydroxy-5-4-methoxyphenyl-4-3,4,5-trimethoxyphenyl methyl, 3-1,3-benzodioxol-5-yl-5-hydroxy-5-4-methoxyphenyl-4-3,4,5-trimethoxybenzyl-2 5h-furanone, 3-2h-1,3-benzodioxol-5-yl-5-hydroxy-5-4-methoxyphenyl-4-3,4,5-trimethoxyphenyl methyl furan-2-one, 3-benzo d 1,3 dioxol-5-yl-5-hydroxy-5-4-methoxyphenyl-4-3,4,5-trimethoxybenzyl furan-2 5h-one, 3-benzo 1,3 dioxol-5-yl-5-hydroxy-5-4-methoxyphenyl-4-3,4,5-trimethoxy-benzyl-5h-furan-2-one, 3-benzo 1,3-dioxol-5-yl-5-hydroxy-5-4-methoxy-phenyl-4-3,4,5-trimethoxy-benzyl-5h-furan-2-one
3-(1,3-benzodioxol-5-yl)-5-hydroxy-5-(4-methoxyphenyl)-4-[(3,4,5-trimethoxyphenyl)methyl]furan-2-one
C28H26O9
PWIPORDFWDZCJG-UHFFFAOYSA-N
9870830
COC1=CC=C(C=C1)C2(C(=C(C(=O)O2)C3=CC4=C(C=C3)OCO4)CC5=CC(=C(C(=C5)OC)OC)OC)O

Specifications

10mg
162256-50-0
chembl8981, 3-1,3-benzodioxol-5-yl-5-hydroxy-5-4-methoxyphenyl-4-3,4,5-trimethoxyphenyl methyl-2 5h-furanone, 3-benzo 1,3 dioxol-5-yl-5-hydroxy-5-4-methoxy-phenyl-4-3,4,5-trimethoxy-benzyl-5h-furan-2-one, 2 5h-furanone,3-1,3-benzodioxol-5-yl-5-hydroxy-5-4-methoxyphenyl-4-3,4,5-trimethoxyphenyl methyl, 3-1,3-benzodioxol-5-yl-5-hydroxy-5-4-methoxyphenyl-4-3,4,5-trimethoxybenzyl-2 5h-furanone, 3-2h-1,3-benzodioxol-5-yl-5-hydroxy-5-4-methoxyphenyl-4-3,4,5-trimethoxyphenyl methyl furan-2-one, 3-benzo d 1,3 dioxol-5-yl-5-hydroxy-5-4-methoxyphenyl-4-3,4,5-trimethoxybenzyl furan-2 5h-one, 3-benzo 1,3 dioxol-5-yl-5-hydroxy-5-4-methoxyphenyl-4-3,4,5-trimethoxy-benzyl-5h-furan-2-one, 3-benzo 1,3-dioxol-5-yl-5-hydroxy-5-4-methoxy-phenyl-4-3,4,5-trimethoxy-benzyl-5h-furan-2-one
COC1=CC=C(C=C1)C2(C(=C(C(=O)O2)C3=CC4=C(C=C3)OCO4)CC5=CC(=C(C(=C5)OC)OC)OC)O
506.507
506.5
CI 1020
C28H26O9
PWIPORDFWDZCJG-UHFFFAOYSA-N
3-(1,3-benzodioxol-5-yl)-5-hydroxy-5-(4-methoxyphenyl)-4-[(3,4,5-trimethoxyphenyl)methyl]furan-2-one
9870830
>98%
Videos
Documents