missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ CHUK (IKK α) Recombinant Human Protein
CHUK (IKK alpha) belongs to the cytokine activated protein complex that is an inhibitor of the NF-kappa B complex.
£1604.00 - £6900.00
Specifications
Accession Number | NP_001269 |
---|---|
For Use With (Application) | Enzyme and Protein Activity Assay, Kinase Assay, Protein Assays and Analysis, Protein Biology |
Formulation | 50mM Tris-HCl pH 7.5, 150mM NaCl, 0.5mM EDTA, 0.02% Triton™ X-100, 2mM DTT, and50% Glycerol |
Molecular Weight (g/mol) | 88.7kDa |
Storage Requirements | Store at -80°C. At first use, aliquot and store at -80°C to avoid multiple freeze-thaws. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15468613
|
Invitrogen™
PR7619B |
100μg |
£1604.00
100µg |
Estimated Shipment: 02-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10056782
|
Invitrogen™
PV4327 |
1mg |
£6900.00
1mg |
Estimated Shipment: 02-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
CHUK (IKK alpha) is a serine/threonine kinase involved in signal transduction and cellular communication.
Specifications
NP_001269 | |
50mM Tris-HCl pH 7.5, 150mM NaCl, 0.5mM EDTA, 0.02% Triton™ X-100, 2mM DTT, and50% Glycerol | |
Store at -80°C. At first use, aliquot and store at -80°C to avoid multiple freeze-thaws. | |
Full Length | |
IKK Kinase Family | |
Recombinant | |
Baculovirus |
Enzyme and Protein Activity Assay, Kinase Assay, Protein Assays and Analysis, Protein Biology | |
88.7kDa | |
RUO - research use only | |
CHUK (IKK α) | |
Human | |
His Tag | |
Frozen |