missing translation for 'onlineSavingsMsg'
Learn More

Chromplete™ Acetonitrile, HPLC/GC, meets ACS/USP analytical requirements, Thermo Scientific™

Catalog Number 15955310
Click to view available options
Quantity:
1 L
2.5 L
4 L
Packaging:
Aluminum Jug
75-05-8
C2H3N
41.05
WEVYAHXRMPXWCK-UHFFFAOYSA-N
methyl cyanide, cyanomethane, ethanenitrile, methanecarbonitrile, ethyl nitrile, methane, cyano, acetonitrile, cyanure de methyl, methylkyanid, mecn
6342
CHEBI:38472
acetonitrile
CC#N
75-05-8
C2H3N
41.05
WEVYAHXRMPXWCK-UHFFFAOYSA-N
methyl cyanide, cyanomethane, ethanenitrile, methanecarbonitrile, ethyl nitrile, methane, cyano, acetonitrile, cyanure de methyl, methylkyanid, mecn
6342
CHEBI:38472
acetonitrile
CC#N

The Chromplete product line from Thermo Scientific includes reagents for common analyses and separations with the consistency and precision you expect. Chromplete Acetonitrile is suitable for HPLC, GC, SFC, TLC, ACS, Spectrophotometry, and meets the analytical requirements of USP.

Chromplete Acetonitrile meets the analytical requirements of USP and is suitable for use in:

  • HPLC coupled with UV, VIS, PDA, ECD, FL, and RI detectors
  • GC- FID, GC- ECD, GC-TCD
  • Pesticide Analysis by GC or HPLC
  • UV Spectrophotometry
  • Preparative chromatography
  • Supercritical Fluid and Thin Layer chromatography
  • Reactions/Analyses requiring ACS compliance
  • Extraction/Concentration Analyses
TRUSTED_SUSTAINABILITY
Melting Point -51°F
Absorbance <0.1 at 195 nm, <0.05 at 200 nm, <0.02 at 210 nm, <0.01 at 220 nm
CAS Min % 0.9995
Color ≤ 5 APHA
UN Number 1648
Formula Weight 41.05
Percent Purity 0.9995
Grade Chromplete
Density 0.775 - 0.78
Boiling Point 180°F
Quantity 2.5 L
Packaging Aluminum Jug
Physical Form Liquid
Refractive Index 1.3405 – 1.3425
Residue after Evaporation ≤ 0.8 ppm
Titratable Acid ≤ 0.008 mEq/g
Titratable Base ≤ 0.0006 mEq/g
Water ≤ 0.01 %
Assay Minimum, GC-FID
Chemical Name or Material Acetonitrile
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Acetonitrile Chromplete
Signal Word
  • Danger
Hazard Category
  • Flammable liquid Category 2
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H302-Harmful if swallowed.
  • H312-Harmful in contact with skin.
  • H319-Causes serious eye irritation.
  • H332-Harmful if inhaled.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'