missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Proteintech ChromoTek Vimentin-Label Atto488
Click to view available options
Quantity:
10 μL
100 μL
Unit Size:
100µL
10µL
Description
- anti-vimentin VHH/ Nanobody conjugated to the fluorophore ATTO488 for IF, & microscopy
Specifications
Specifications
| Clonality | Monoclonal |
| Quantity | 100 μL |
| For Use With (Application) | Immunofluorescence (IF), Immunocytochemistry (ICC) |
| Conjugate | ATTO488 |
| Target Specificity | Vimentin-Label specifically binds to the vimentin intermediate filament protein. Validated on canine MDCK cells, rodent BHK cells and on human A549 cells upon stimulation with TGFβ. |
| Content And Storage | Shipped at ambient temperature. Upon receipt store at 4°C. Do not freeze. Protect from light. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction