missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Proteintech ChromoTek Spot-Label™ Alexa Fluor™ 647
Click to view available options
Quantity:
10 μL
50 μL
Unit Size:
10µL
50µL
Description
- anti-Spot-Tag VHH/ Nanobody conjugated to fluorescent dye for immunofluorescence, microscopy, and immunoblotting of Spot-tagged proteins
Specifications
Specifications
| Clonality | Monoclonal |
| Quantity | 50 μL |
| For Use With (Application) | Immunofluorescence (IF), Western Blot |
| Conjugate | Alexa Fluor™ 647 |
| Target Specificity | Spot-Tag (PDRVRAVSHWSS) |
| Content And Storage | Shipped at ambient temperature. Upon receipt store at +4°C / 40°F.Stable for 6 months. Do not freeze. Protect from light. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction