Learn More
Proteintech ChromoTek RFP-Trap™ Magnetic Particles M-270 Kit
Click to view available options
Quantity:
20 Reactions
Unit Size:
20 reactions
Description
- RFP-Trap™ Magnetic Particles M-270 for immunoprecipitation (IP) of RFP-tagged proteins
- RFP-Trap Magnetic Particles M-270 is highly recommended, when very large proteins/complexes are investigated, and magnetic separation is needed for IP
- Consists of a RFP VHH/ Nanobody coupled to Magnetic Particles M-270
Specifications
Specifications
| Quantity | 20 Reactions |
| Sufficient For | 20 Reactions |