Learn More
Proteintech ChromoTek PARP1-Trap Agarose Beads Kit
Click to view available options
Quantity:
20 Reactions
Unit Size:
20 reactions
Description
- PARP1-Trap Agarose is an affinity reagent for IP of human PARP1
- Consists of a PARP1 Nanobody/ VHH, which is coupled to agarose beads
Specifications
Specifications
| Quantity | 20 Reactions |
| Sufficient For | 20 Reactions |