Learn More
Proteintech ChromoTek Myc-Trap™ Agarose Kit
Product Code. 17373573
Click to view available options
Quantity:
20 Reactions
Unit Size:
20 reactions
Product Code. 17373573
Brand: Proteintech ytak20
Kit including 20 reactions of Myc-Trap™ Agarose and lysis, dilution, wash, and elution buffers.
  • Myc-Trap™ Agarose is an affinity reagent for IP and purification of Myc-tagged proteins
  • Consists of a Myc Nanobody/ VHH coupled to agarose beads
Specifications
Quantity 20 Reactions
Sufficient For 20 Reactions