Learn More
Proteintech ChromoTek MK2-Trap Agarose Kit
Product Code. 17383383
Click to view available options
Quantity:
20 Reactions
Unit Size:
20 reactions
Product Code. 17383383
Brand: Proteintech mtak20
Kit including 20 reactions of MK2-Trap Agarose and lysis, dilution, wash, and elution buffers.
  • ChromoTek’s MK2-Trap Agarose kit are affinity beads for IP of MK2-fusion proteins
  • Comprises a MK2 Nanobody/ VHH conjugated to agarose beads
Specifications
Quantity 20 Reactions
Sufficient For 20 Reactions