Learn More
Proteintech ChromoTek iST TurboGFP-Trap Kit
Click to view available options
Quantity:
8 Reactions
Unit Size:
8 reactions
Description
- Kit for IP/ Co-IP of TurboGFP-tagged proteins & sample preparation for mass spectrometry (MS)
Specifications
Specifications
| Quantity | 8 Reactions |
| Sufficient For | 8 Reactions |