missing translation for 'onlineSavingsMsg'
Learn More

Proteintech ChromoTek GST-Trap Agarose

Product Code. 17383503
Click to view available options
Sufficient For:
10 Reactions
100 Reactions
20 Reactions
200 Reactions
400 Reactions
Unit Size:
10 reactions
100 reactions
20 reactions
200 reactions
400 reactions
This item is not returnable. View return policy

Product Code. 17383503

Brand: Proteintech sta400

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

GST-Trap is an affinity resin for IP of GST fusion proteins.

  • Consists of a GST Nanobody/ VHH coupled to agarose bead

Specifications

Content And Storage Upon receipt store at 4°C. Do not freeze.
For Use With (Application) IP, CoIP, ChIP, RIP
Target Glutathione S-Transferase (GST) from Schistosoma japonicum
Label or Dye Agarose
Species Alpaca
Sufficient For 400 Reactions
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.