missing translation for 'onlineSavingsMsg'
Learn More

Chromone-3-carbonitrile, 97%, Thermo Scientific™

Catalog Number p-7022776
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
50743-17-4
C10H5NO2
171.16
MFCD00052604
SFWNPLLGXKJESA-UHFFFAOYSA-N
3-cyanochromone, chromone-3-carbonitrile, 4-oxo-4h-chromene-3-carbonitrile, 4-oxo-4h-1-benzopyran-3-carbonitrile, acmc-20an5h, maybridge1_006148, 4-oxo-chromene-3-carbonitrile, 3-cyano-4h-1-benzopyran-4-one, 4-oxo-1-benzopyran-3-carbonitrile
521298
O=C1C(=COC2=CC=CC=C12)C#N
This item is not returnable. View return policy
50743-17-4
C10H5NO2
171.16
MFCD00052604
SFWNPLLGXKJESA-UHFFFAOYSA-N
3-cyanochromone, chromone-3-carbonitrile, 4-oxo-4h-chromene-3-carbonitrile, 4-oxo-4h-1-benzopyran-3-carbonitrile, acmc-20an5h, maybridge1_006148, 4-oxo-chromene-3-carbonitrile, 3-cyano-4h-1-benzopyran-4-one, 4-oxo-1-benzopyran-3-carbonitrile
521298
O=C1C(=COC2=CC=CC=C12)C#N

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Melting Point 173°C to 177°C
Quantity 1 g
UN Number UN3439
IUPAC Name 4-oxo-4H-chromene-3-carbonitrile
Formula Weight 171.16
Percent Purity 97%
Chemical Name or Material Chromone-3-carbonitrile
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'