missing translation for 'onlineSavingsMsg'
Learn More

Chromium Standard For ICP, 1000μg/mL in 2% HNO3, Assurance Grade, SPEX CertiPrep™

Catalog Number p-5533051
Click to view available options
Quantity:
125 mL
250 mL
500 mL
Packaging:
Triple-leached LDPE bottle
This item is not returnable. View return policy
7789-09-5, 7732-18-5
Cr2H8N2O7
252.06
JOSWYUNQBRPBDN-UHFFFAOYSA-P
24600
diazanium;oxido-(oxido(dioxo)chromio)oxy-dioxochromium
[NH4+].[NH4+].[O-][Cr](=O)(=O)O[Cr](=O)(=O)[O-]
This item is not returnable. View return policy
7789-09-5, 7732-18-5
Cr2H8N2O7
252.06
JOSWYUNQBRPBDN-UHFFFAOYSA-P
24600
diazanium;oxido-(oxido(dioxo)chromio)oxy-dioxochromium
[NH4+].[NH4+].[O-][Cr](=O)(=O)O[Cr](=O)(=O)[O-]

Includes

NIST Traceable Certificate of Analysis accredited to ISO/IEC 17025 and ISO/IEC Guide 34, Safety Data Sheet

ISO/IEC Guide 34 Certified, NIST-Traceable Certified Reference Material

Inorganic Standard manufactured for AA, ICP, ICP/MS, IC, XRF, and other analytical instrumentation

AnalyteConcentration
Cr1000μg/mL

Certifications

Accredited by A2LA for ISO/IEC 17025 and ISO/IEC Guide 34 and certified by UL-DQS to ISO 9001

Compliance

GHS/OSHA Complaint

Grade Assurance
Packaging Triple-leached LDPE bottle
Certificate of Analysis Assurance Certificate of Analysis reports up to 39 elements at ppb levels.
Solvent or Matrix 2% HNO3
Quantity 125 mL
For Use With (Equipment) AA, ICP, ICP/MS, IC, XRF, and Other Analytical Instrumentation
Concentration or Composition Notes Up to 68 elements are scanned with found values for ICP and ICP-MS standards. The found value of the trace metallic impurities is reported on the Certificate of Analysis.
Chemical Name or Material Chromium Standard

Product intended for laboratory use only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'