missing translation for 'onlineSavingsMsg'
Learn More

Chromium sputtering target, 50.8mm (2.0in) dia x 3.18mm (0.125in) thick, 99.95% (metals basis), Thermo Scientific Chemicals

Catalog Number 15474785
Click to view available options
Quantity:
1 Ea.
This item is not returnable. View return policy
This item is not returnable. View return policy

 

Chromium sputtering target is commonly used for thin-film deposition, etching and analytical techniques. And It is also used as Magnetron sputtering source.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Quantity 1 Ea.
CAS 7440-47-3
Odor Odorless
Molecular Formula Cr
MDL Number MFCD00010944
Synonym chrom, chrome, cromo, metal, chrome french, compounds, chrom german, chromium, elemental, chromium, metal, ccris 159
Solubility Information Insoluble in water.
InChI Key VYZAMTAEIAYCRO-UHFFFAOYSA-N
SMILES [Cr]
IUPAC Name chromium
Molecular Weight (g/mol) 52.00
PubChem CID 23976
ChEBI CHEBI:28073
Percent Purity 99.95%
Assay (metals basis)
Chemical Name or Material Chromium sputtering target, 50.8mm (2.0 in.) dia. x 3.18mm (0.125 in.) thick
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'