missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Chromium(III) telluride, 99.5% (metals basis), Thermo Scientific Chemicals
42.00 GBP valid until 2024-03-29
Use promo code "21615" to get your promotional price.
Use promo code "21615" to get your promotional price.
Click to view available options
Quantity:
5 g
25 g
Description
Chromium telluride is used in a number of industrial applications including infrared detection and imaging.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Melting Point | ∼1300°C |
Color | Gray to Black |
Physical Form | Powder |
Quantity | 5 g |
Solubility Information | Insoluble in water. |
Formula Weight | 486.79 |
Percent Purity | 99.5% |
Odor | Odorless |
Density | 7.0 g/mL |
Assay | (metals basis) |
Show More |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'