missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Chromium(III) oxide sputtering target, 76.2mm (3.0 in.) dia. x 3.18mm (0.125 in.) thick, 99.8% (metals basis), Thermo Scientific™
Click to view available options
Quantity:
1 Ea.
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifications
Boiling Point | 3000°C |
Melting Point | 2439°C |
CAS | 1308-38-9 |
Color | Green |
Quantity | 1 Ea. |
Molecular Formula | Cr2O3 |
MDL Number | MFCD00010949 |
Synonym | chromium iii oxide, chromia, dichromium trioxide, chromium sesquioxide, chrome green, green chromium oxide, chromium 3+ oxide, green cinnabar, chrome ochre, chrome oxide |
InChI Key | UOUJSJZBMCDAEU-UHFFFAOYSA-N |
SMILES | [O--].[O--].[O--].[Cr+3].[Cr+3] |
Show More |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'