missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Chromium(III) nitrate nonahydrate, 99.99% min (metals basis), Thermo Scientific Chemicals
29.86 GBP valid until 2024-03-29
Use promo code "21615" to get your promotional price.
Use promo code "21615" to get your promotional price.
Click to view available options
Quantity:
5 g
25 g
100 g
Description
Chromium nitrate is used in the manufacture of alkali metal-free catalysts and in preservation. It is used in the preparation of chrome catalysts, in textile printing operations, and as a corrosion inhibitor. Chromium(III) nitrate as a source of chromium is added to food supplements for nutritional purposes .
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Melting Point | 60°C |
Color | Purple |
Physical Form | Crystalline |
Quantity | 5 g |
UN Number | UN2720 |
Merck Index | 14,2226 |
Solubility Information | Soluble in water and alcohol. |
Formula Weight | 400.14 (238.01 Anhydrous) |
Percent Purity | ≥99.99% |
Odor | Odorless |
Show More |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'