missing translation for 'onlineSavingsMsg'
Learn More

Chromium(III) chloride tetrahydrofuran complex (1:3), 98%, ACROS Organics™

Brand:  Acros Organics 389850100

Code : 29

Additional Details : CAS Number : 10170-68-0 Weight : 0.06000kg Transport : UN number : 1325 Chem class : 4.1 Pack group : II


Packaging Glass bottle
Quantity 10g

 View more versions of this product

Product Code. 10702984

  • £100.66 / 10g

In stock, ready to ship: 2

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Chemical Identifiers

10170-68-0
374.67
CYOMBOLDXZUMBU-UHFFFAOYSA-K
11100829
C1CCOC1.C1CCOC1.C1CCOC1.Cl[Cr](Cl)Cl
C12H24Cl3CrO3
MFCD00013222
chromium iii chloride tetrahydrofuran complex 1:3, chromium iii chloride tetrahydrofuran adduct, crcl3 thf 3, crcl3 3thf, chromium iii chloride tetrahydrofuran complex, chromium chloride; tris tetrahydrofuran, chromium iii chloride complex 1:3 tetrahydrofuran
oxolane;trichlorochromium

Specifications

Chromium(III) chloride tetrahydrofuran complex (1:3)
98%
MFCD00013222
CYOMBOLDXZUMBU-UHFFFAOYSA-K
oxolane;trichlorochromium
11100829
Crystalline Powder
97.5% min.
Glass bottle
143.0°C
10170-68-0
C12H24Cl3CrO3
chromium iii chloride tetrahydrofuran complex 1:3, chromium iii chloride tetrahydrofuran adduct, crcl3 thf 3, crcl3 3thf, chromium iii chloride tetrahydrofuran complex, chromium chloride; tris tetrahydrofuran, chromium iii chloride complex 1:3 tetrahydrofuran
C1CCOC1.C1CCOC1.C1CCOC1.Cl[Cr](Cl)Cl
374.67
374.67
98%
Authentic
Purple
10g
Videos
Safety and Handling

Safety and Handling

Product Identifier
  • Chromium(III) chloride tetrahydrofuran complex (1:3)

Signal Word
  • Warning

Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Flammable solid Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3

Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H302-Harmful if swallowed.
  • H228-Flammable solid.
  • EUH019-May form explosive peroxides.

Precautionary Statement
  • P243-Take precautionary measures against static discharge.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P381-Eliminate all ignition sources if safe to do so.

SDS Documents