missing translation for 'onlineSavingsMsg'
Learn More

Chromium(III) chloride hexahydrate, 99.5% min, Thermo Scientific Chemicals

Catalog Number p-7051852
Click to view available options
Quantity:
100 g
500 g
10060-12-5
Cl3CrH12O6
266.44
MFCD00149660
LJAOOBNHPFKCDR-UHFFFAOYSA-K
chromium iii chloride hexahydrate, chromic chloride hexahydrate, hexaaquachromium chloride, chromium trichloride hexahydrate, trichlorochromium hexahydrate, chromium chloride hexahydrate, hexaaquachromium iii chloride, chrom iii-chlorid, chromium 3+ chloride hexahydrate, chlorid chromity hexahydrat czech
104957
CHEBI:53442
O.O.O.O.O.O.[Cl-].[Cl-].[Cl-].[Cr+3]
10060-12-5
Cl3CrH12O6
266.44
MFCD00149660
LJAOOBNHPFKCDR-UHFFFAOYSA-K
chromium iii chloride hexahydrate, chromic chloride hexahydrate, hexaaquachromium chloride, chromium trichloride hexahydrate, trichlorochromium hexahydrate, chromium chloride hexahydrate, hexaaquachromium iii chloride, chrom iii-chlorid, chromium 3+ chloride hexahydrate, chlorid chromity hexahydrat czech
104957
CHEBI:53442
O.O.O.O.O.O.[Cl-].[Cl-].[Cl-].[Cr+3]

Chromium(III) chloride hexahydrate is used as a catalyst, as a mordant in printing and dyeing industry. It is involved in the preparation of anhydrous tetrahydrofuran complex of chromium(III) chloride by reacting with trimethylsilylchloride. It finds application in the manufacturing of a variety of chromium-containing pigment in Paint industry and other chromium salts in the chemical industry.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point 83°C
Color Black to Green
Physical Form Crystalline
Quantity 100 g
UN Number UN3260
Merck Index 14,2222
Solubility Information Slightly soluble in acetone. Soluble in water,alcohol. Insoluble in ether
IUPAC Name trichlorochromium;hexahydrate
Formula Weight 266.45 (158.35 Anhydrous)
Percent Purity ≤99.5%
Odor Odorless
Sensitivity Hygroscopic
Density 1.76 g/mL
Chemical Name or Material Chromium(III) chloride hexahydrate
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'