missing translation for 'onlineSavingsMsg'
Learn More

Chromium(II) chloride, 99.9% (metals basis), Thermo Scientific Chemicals

Catalog Number p-7052309
Click to view available options
Quantity:
1 g
5 g
25 g
This item is not returnable. View return policy
10049-05-5
MFCD00010947
Chromous chloride
This item is not returnable. View return policy
10049-05-5
MFCD00010947
Chromous chloride

Chromium(II) chloride is used in the condensation of aldehydes with trisubstituted chloroalkenes as well as in the synthesis of a highly active precatalyst for ethylene oligimerization. Further uses of chromium(II) chloride include the conversion of terminal olefins into trans-iodocyclopropanes (in conjunction with iodoform and tetraethyl ethylenediamine) with excellent results; generating bis(indenyl)chromium dimers and other various related products.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point 824°C
Color Gray
Physical Form Powder
Quantity 1 g
Linear Formula CrCl2
UN Number UN3260
Merck Index 14,2244
Solubility Information Soluble in water. Insoluble in alcohol and ether.
Formula Weight 122.9
Percent Purity 99.9%
Odor Odorless
Sensitivity Air and moisture sensitive
Density 2.878 g/mL
Assay (metals basis)
Chemical Name or Material Chromium(II) chloride
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'