Learn More
Chromium(II) chloride, 99.9% (metals basis), Thermo Scientific Chemicals
Use promo code "21615" to get your promotional price.
Description
Chromium(II) chloride is used in the condensation of aldehydes with trisubstituted chloroalkenes as well as in the synthesis of a highly active precatalyst for ethylene oligimerization. Further uses of chromium(II) chloride include the conversion of terminal olefins into trans-iodocyclopropanes (in conjunction with iodoform and tetraethyl ethylenediamine) with excellent results; generating bis(indenyl)chromium dimers and other various related products.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Melting Point | 824°C |
Color | Gray |
Physical Form | Powder |
Quantity | 25 g |
Linear Formula | CrCl2 |
UN Number | UN3260 |
Merck Index | 14,2244 |
Solubility Information | Soluble in water. Insoluble in alcohol and ether. |
Formula Weight | 122.9 |
Percent Purity | 99.9% |
Show More |
RUO – Research Use Only