Learn More
Alfa Aesar™ Chromium(II) chloride, 99.9% (metals basis)
Brand: Alfa Aesar™ 047176.06
Code : NEW
Additional Details : CAS Number : 10049-05-5 Weight : 0.00500kg Transport : UN number : 3260 Chem class : 1a Pack group : III
Quantity | 5g |
---|
Chemical Identifiers
Chromium(II) chloride | |
CrCl2 | |
MFCD00010947 | |
14,2244 |
10049-05-5 | |
122.9 | |
UN3260 | |
Chromous chloride |
Specifications
Chromium(II) chloride | |
Powder | |
(metals basis) | |
122.9 | |
UN3260 | |
Chromous chloride | |
−80 Mesh | |
Air & Moisture Sensitive | |
824°C |
10049-05-5 | |
99.9% | |
CrCl2 | |
MFCD00010947 | |
14,2244 | |
2.878g/mL | |
Odorless | |
Gray | |
5g |