missing translation for 'onlineSavingsMsg'
Learn More

Chromium(II) chloride, 90%, Thermo Scientific Chemicals

Catalog Number 10521861
Click to view available options
Quantity:
5 g
10 g
Packaging:
Glass bottle
This item is not returnable. View return policy
10049-05-5
Cl2Cr
122.9
MFCD00010947
Chromous chloride
This item is not returnable. View return policy
10049-05-5
Cl2Cr
122.9
MFCD00010947
Chromous chloride

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Reduction reagent
TRUSTED_SUSTAINABILITY
Melting Point 824.0°C
Color Gray to Green
Physical Form Powder
Quantity 10 g
Assay Percent Range 88% min. (Argentometry)
Linear Formula CrCl2
Fieser 01,149; 02,76; 03,60; 04,144; 07,73; 09,511; 12,136; 13,84; 14,94; 16,93; 17,84
Merck Index 15, 2248
Solubility Information Solubility in water: may decompose. Other solubilities: insoluble in alcohol and ether
Formula Weight 122.9
Percent Purity 90%
Packaging Glass bottle
Chemical Name or Material Chromium(II) chloride, 90%
Show More Show Less
compliance-icons
Product Identifier
  • Chromium(II) chloride
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P332+P313-If skin irritation occurs: Get medical advice/attention.
  • P337+P313-If eye irritation persists: Get medical advice/attention.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'