missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ Chromium flakes, 2-12mm (0.08-0.47in), 99.996% (metals basis), Oxygen <100ppm, Nitrogen <50ppm

Brand:  Alfa Aesar™ 044005.09

Code : NEW

Additional Details : CAS Number : 7440-47-3 Weight : 0.01000kg

Quantity 10g

 View more versions of this product

Product Code. 11345078

  • £59.25 / 10g


Estimated Shipment date
from Supplier 25-05-2018
Add to basket

Specifications

Chemical Identifiers

Chromium flakes
7440-47-3
2 to 12mm (0.08 to 0.47 in.), Oxygen <100ppm, Nitrogen <50ppm
MFCD00010944

Specifications

Chromium flakes
2 to 12mm (0.08 to 0.47 in.), Oxygen <100ppm, Nitrogen <50ppm
Flakes
(metals basis)
Odorless
10g
7440-47-3
99.996%
MFCD00010944
Videos
SDS