Learn More
Retsch™ Chrome Steel Comfort Grinding Jars
Use these grinding jars with Retsch™ ball mills. Retsch™ Chrome Steel Comfort Grinding Jars feature an O-type sealing for safe operation.
Brand:  Retsch™ 01.462.0229
Code : 50
Additional Details : Weight : 4.10000kg
 View more versions of this product
Product Code. 10789944
  • £1728.00 / Each
Estimated Shipment
01-03-2018
Add to basket
Description
Description
  • Available in a variety of sizes
  • Optional aeration covers
  • Pressure tight safety closure devices
Specifications
Specifications
500mL
Chrome Steel
For PM 100, PM 400
Comfort Grinding Jar
Videos
View accessory products