Learn More
Thermo Scientific Maybridge Chroman-8-sulfonyl chloride, 1g, 97%
Brand: Thermo Scientific Maybridge CC76103DA
Chroman-8-sulfonyl chloride, 1g, 97%
Code : 31
Additional Details : CAS Number : 1048970-15-5 Weight : 0.05100kg Transport : UN number : 3261 Chem class : 8 Pack group : III