Learn More
Thermo Scientific Maybridge Chroman-8-sulfonyl chloride, 5g, 97%
Brand: Thermo Scientific Maybridge CC76103DE
Chroman-8-sulfonyl chloride, 5g, 97%
Code : 31
Additional Details : CAS Number : 1048970-15-5 Weight : 0.05500kg
Brand: Thermo Scientific Maybridge CC76103DE
Chroman-8-sulfonyl chloride, 5g, 97%
Code : 31
Additional Details : CAS Number : 1048970-15-5 Weight : 0.05500kg