Learn More
Thermo Scientific Maybridge Chroman-6-sulfonyl chloride, 1g, 95%
Brand: Thermo Scientific Maybridge CC32803DA
Chroman-6-sulfonyl chloride, 1g, 95%
Code : 31
Additional Details : CAS Number : 946409-11-6 Weight : 0.05100kg
Brand: Thermo Scientific Maybridge CC32803DA
Chroman-6-sulfonyl chloride, 1g, 95%
Code : 31
Additional Details : CAS Number : 946409-11-6 Weight : 0.05100kg