Macherey Nagel™ Chromafix™ RP Development Kit I
Overview
Brand: Macherey Nagel™
Manufacturer Part Number: 731886
UNSPSC: 41115700
Code: 50
Additional Details:
Additional Details: Weight: 0.01000kg
Product Code. 10585381
Quantity Price
1 £ 91.45 / Each
Estimated Shipment
11-04-2017
Add to basket
Description and Specification
Specification
Product Type Chromafix RP Development Kit I
Includes C18, C18 ec, C18 Hydra, C8, HR-P, HR-X Cartridges