Learn More
Macherey-Nagel™ Chromafix™ HR-XCW Cartridge
Brand: Macherey-Nagel™ 731776
Code : 50
Additional Details : Weight : 0.10000kg
Specifications
Spherical PS/DVB Copolymer | |
Basic Compounds Like Quaternary Amines, Basic Active Ingredients From Strongly Matrix-Contaminated Samples | |
Weak carboxylic acid modified polymeric cation exchanger | |
50 to 60 Å | |
850 m2/g |
500 mg | |
45 or 85 μm | |
Chromafix HR-XCW Cartridge | |
50 Pack |