missing translation for 'onlineSavingsMsg'
Learn More

Macherey-Nagel™ CHROMABOND™ XTR Polypropylene SPE Columns

Designed for consistently high quality from batch to batch. Macherey-Nagel™ CHROMABOND™ XTR Polypropylene SPE Columns include Kieselguhr phase for liquid-liquid extractions. Columns feature a large pore size and high pore volume for use with highly viscous aqueous solutions. Volumes from 1–45mL.

Brand:  Macherey-Nagel™ 730502

Code : 50

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 10690532

  • £92.95 / Pack of 50
Estimated Shipment: 14-06-2024
to see stock.
Add to basket

Alert:

Eluents with too high alcohol contents cause an increase in volume of the aqueous phase on the CHROMABOND™ XTR. Here the column could be overloaded and the aqueous phase displaced from the column. In this case, a greater capacity column should be used.


Description

Description

Polypropylene SPE columns with a base material of coarse-grained kieselguhr (also known as diatomaceous earth, hydromatrix, celite), with a large pore size and high pore volume. The pH value of aqueous solutions can be altered on the column, from 1–13, which enables elution of different compounds of a sample under optimized conditions.

Application:
For liquid-liquid extraction of highly viscous aqueous solutions without use of a separation funnel. Columns feature high water loadability without breakthrough during elution with organic solvents. They are also suited for removing small amounts of water from solvents which are not miscible with water. 

  • Physiological fluids (blood, plasma, and serum) in clinical chemistry
  • Dyes in textiles
  • Environmental and food analysis

Advantages:

  • Fast, reproducible and economical
  • Simultaneous preparation of several samples
  • No problems with phase separation—no formation of emulsions
  • High recovery rates
  • Saving of time and solvents
  • Organic solutions do not need to be dried after separation
Specifications

Specifications

3 mL
500 mg
Chromabond XTR
Polypropylene
Liquid-Liquid Extraction
Chromabond XTR Column
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers