missing translation for 'onlineSavingsMsg'
Learn More

Macherey Nagel™ Chromabond™ PCA Polypropylene SPE Column

Brand:  Macherey Nagel™ 730483

UNSPSC : 23151816

Code : 50

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 10649001

  • £162.00 / Pack of 30


Call for latest availability
+44 (0)1509 555 500
Add to basket

Specifications Description & Specifications

Specifications

60μ
6mL
45μm
Chromabond® PCA
Silica
Silica
SPE
SPE Column
For SPE
500m2/g