missing translation for 'onlineSavingsMsg'
Learn More

Macherey Nagel™ Chromabond™ HR-XCW Polypropylene SPE Column

Brand:  Macherey Nagel™ 730741

UNSPSC : 23151816

Code : 50

Additional Details : Weight : 0.20000kg

 View more versions of this product

Product Code. 12728212

  • £203.00 / Pack of 30


Call for latest availability
+44 (0)1509 555 500
Add to basket

Specifications

Specifications

Spherical PS/DVB Copolymer
SPE
SPE Column
850m2/g
85μm
3mL
50 to 60μ
PS/DVB Copolymer
Chromabond HR-XCW