missing translation for 'onlineSavingsMsg'
Learn More

Macherey Nagel™ Chromabond™ Easy Polypropylene SPE Column

Brand:  Macherey Nagel™ 730758

UNSPSC : 23151816

Code : 50

Additional Details : Weight : 0.20000kg

 View more versions of this product

Product Code. 12887821

  • £362.00 / Each


Call for latest availability
+44 (0)1509 555 500
Add to basket

Specifications Description & Specifications

Specifications

15mL
For SPE
SPE Column
Chromabond Easy
SPE
50μ
650 to 700m2/g
80μm