missing translation for 'onlineSavingsMsg'
Learn More

Macherey-Nagel™ Chromabond™ Easy Polypropylene SPE Column

Brand:  Macherey-Nagel™ 730754

Code : 50

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 10766522

  • £135.00 / Pack of 30


Call for latest availability
+44 (0)1509 555 500
Add to basket

Specifications

Specifications

6mL
For SPE
SPE Column
Chromabond Easy
SPE
50μ
650 to 700m2/g
80μm
Videos